Assay Details
Gene Symbol: Tmem9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105953.2 | NP_001099423.2 | 5 - 6 | 484 | 94 | |
NM_001277263.1 | NP_001264192.1 | 5 - 6 | 561 | 94 | ||
GenBank mRNA | BC169086.1 | - | 4 - 5 | 451 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
