Assay Details
Gene Symbol: Uhmk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017293.1 | NP_058989.1 | 6 - 7 | 1185 | 97 | |
GenBank mRNA | U70372.2 | - | 6 - 7 | 1185 | 97 | |
X98374.1 | - | 6 - 7 | 1062 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
