Assay Details
Gene Symbol: Dstyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199463.2 | NP_955750.1 | 11 - 12 | 2528 | 77 | |
XM_006249774.2 | XP_006249836.1 | 11 - 12 | 2378 | 77 | ||
GenBank mRNA | AY208851.1 | - | 11 - 12 | 2459 | 77 | |
AY429675.1 | - | 11 - 12 | 2528 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
