Assay Details
Gene Symbol: Dstyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199463.2 | NP_955750.1 | 3 - 4 | 1382 | 68 | |
XM_006249774.2 | XP_006249836.1 | 3 - 4 | 1232 | 68 | ||
GenBank mRNA | AY208851.1 | - | 3 - 4 | 1313 | 68 | |
AY429675.1 | - | 3 - 4 | 1382 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
