Assay Details
Gene Symbol: Cd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022269.2 | NP_071605.2 | 7 - 8 | 1100 | 64 | |
GenBank mRNA | AB026901.1 | - | 7 - 8 | 1044 | 64 | |
AB026903.1 | - | 7 - 8 | 1044 | 64 | ||
AB026904.1 | - | 6 - 7 | 893 | 64 | ||
BC061869.1 | - | 7 - 8 | 1064 | 5598916 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
