Assay Details
Gene Symbol: Cd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249723.2 | XP_006249785.1 | 10 - 11 | 1182 | 144 | |
GenBank mRNA | AB026904.1 | - | 8 - 9 | 992 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
