Assay Details
Gene Symbol: Mapkapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178102.2 | NP_835203.1 | 4 - 5 | 745 | 85 | |
GenBank mRNA | AY197741.1 | - | 4 - 5 | 745 | 85 | |
BC062048.1 | - | 4 - 5 | 711 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
