Assay Details
Gene Symbol: Pigr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012723.4 | NP_036855.3 | 118 | 77 | ||
GenBank mRNA | FQ104960.1 | - | 1 - 2 | 118 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_012723.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
