Assay Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153821.1 | NP_722543.1 | 1 - 2 | 564 | 102 | |
XM_006250151.2 | XP_006250213.1 | 1 - 2 | 1302 | 102 | ||
GenBank mRNA | AY730764.1 | - | 1 - 2 | 288 | 102 | |
S82911.1 | - | 1 - 2 | 564 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
