Assay Details
Gene Symbol: Uap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191930.1 | NP_001178859.1 | 1 - 2 | 164 | 123 | |
XM_006250222.1 | XP_006250284.1 | 1 - 2 | 159 | 123 | ||
XM_006250223.1 | XP_006250285.1 | 1 - 2 | 159 | 123 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001191930.1, XM_006250222.1, XM_006250223.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
