Assay Details
Gene Symbol: Uap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250222.1 | XP_006250284.1 | 9 - 10 | 1617 | 149 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
