Assay Details
Gene Symbol: Abl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107186.1 | NP_001100656.1 | 13 - 14 | 3486 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
phagocytosis
substrate-dependent cell migration, cell extension
actin filament organization
epidermal growth factor receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
learning
biological_process
visual learning
positive regulation of phospholipase C activity
positive regulation of neuron projection development
neuron remodeling
cell migration
peptidyl-tyrosine phosphorylation
cerebellum morphogenesis
negative regulation of cell-cell adhesion
actin cytoskeleton organization
cell differentiation
positive regulation of protein binding
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
cellular protein localization
negative regulation of Rho protein signal transduction
exploration behavior
peptidyl-tyrosine autophosphorylation
regulation of cell population proliferation
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
alpha-beta T cell differentiation
platelet-derived growth factor receptor signaling pathway
dendrite morphogenesis
neuromuscular process controlling balance
actin filament bundle assembly
positive regulation of oxidoreductase activity
Bergmann glial cell differentiation
positive regulation of ERK1 and ERK2 cascade
cellular response to retinoic acid
dendritic spine maintenance
regulation of extracellular matrix organization
positive regulation of Wnt signaling pathway, planar cell polarity pathway
negative regulation of endothelial cell apoptotic process
magnesium ion binding
molecular_function
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
signaling receptor binding
ATP binding
manganese ion binding
protein phosphorylation
phagocytosis
substrate-dependent cell migration, cell extension
actin filament organization
epidermal growth factor receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
learning
biological_process
visual learning
positive regulation of phospholipase C activity
positive regulation of neuron projection development
neuron remodeling
cell migration
peptidyl-tyrosine phosphorylation
cerebellum morphogenesis
negative regulation of cell-cell adhesion
actin cytoskeleton organization
cell differentiation
positive regulation of protein binding
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
cellular protein localization
negative regulation of Rho protein signal transduction
exploration behavior
peptidyl-tyrosine autophosphorylation
regulation of cell population proliferation
regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
alpha-beta T cell differentiation
platelet-derived growth factor receptor signaling pathway
dendrite morphogenesis
neuromuscular process controlling balance
actin filament bundle assembly
positive regulation of oxidoreductase activity
Bergmann glial cell differentiation
positive regulation of ERK1 and ERK2 cascade
cellular response to retinoic acid
dendritic spine maintenance
regulation of extracellular matrix organization
positive regulation of Wnt signaling pathway, planar cell polarity pathway
negative regulation of endothelial cell apoptotic process
magnesium ion binding
molecular_function
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
signaling receptor binding
ATP binding
manganese ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P114331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58792 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P114331 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P114331 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P114331 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P114331 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16072 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59742 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17430 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77447 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25571 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100713 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7369 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95807 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14824 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10764858 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764855 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764857 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764856 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10764852 | - | - |
Affymetrix Rat Clariom S Assay | PSR1300003794.rn.1 | TC1300000552.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10764852 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21529680 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351581 | - | - |
