Assay Details
Gene Symbol: Abl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107186.1 | NP_001100656.1 | 4 - 5 | 688 | 117 | |
XM_006250046.2 | XP_006250108.1 | 4 - 5 | 996 | 117 | ||
XM_006250047.2 | XP_006250109.1 | 4 - 5 | 996 | 117 | ||
XM_006250048.2 | XP_006250110.1 | 4 - 5 | 1667 | 117 | ||
XM_006250049.2 | XP_006250111.1 | 4 - 5 | 1666 | 117 | ||
XM_006250050.2 | XP_006250112.1 | 3 - 4 | 933 | 117 | ||
XM_006250051.2 | XP_006250113.1 | 3 - 4 | 933 | 117 | ||
XM_006250052.2 | XP_006250114.1 | 4 - 5 | 996 | 117 | ||
XM_006250053.2 | XP_006250115.1 | 4 - 5 | 995 | 117 | ||
XM_006250054.2 | XP_006250116.1 | 4 - 5 | 1665 | 117 | ||
XM_006250055.2 | XP_006250117.1 | 3 - 4 | 932 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
