Assay Details
Gene Symbol: Abl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107186.1 | NP_001100656.1 | 5 - 6 | 950 | 79 | |
XM_006250046.2 | XP_006250108.1 | 5 - 6 | 1258 | 79 | ||
XM_006250047.2 | XP_006250109.1 | 5 - 6 | 1258 | 79 | ||
XM_006250048.2 | XP_006250110.1 | 5 - 6 | 1929 | 79 | ||
XM_006250049.2 | XP_006250111.1 | 5 - 6 | 1928 | 79 | ||
XM_006250050.2 | XP_006250112.1 | 4 - 5 | 1195 | 79 | ||
XM_006250051.2 | XP_006250113.1 | 4 - 5 | 1195 | 79 | ||
XM_006250052.2 | XP_006250114.1 | 5 - 6 | 1258 | 79 | ||
XM_006250053.2 | XP_006250115.1 | 5 - 6 | 1257 | 79 | ||
XM_006250054.2 | XP_006250116.1 | 5 - 6 | 1927 | 79 | ||
XM_006250055.2 | XP_006250117.1 | 4 - 5 | 1194 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
