Assay Details
Gene Symbol: Rbbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107174.1 | NP_001100644.1 | 1 - 2 | 385 | 78 | |
XM_006249775.2 | XP_006249837.1 | 1 - 2 | 173 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001107174.1,XM_006249775.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
