Assay Details
Gene Symbol: Rbbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107174.1 | NP_001100644.1 | 9 - 10 | 1346 | 142 | |
XM_006249775.2 | XP_006249837.1 | 9 - 10 | 1134 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
