Assay Details
Gene Symbol: Mdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012026.1 | NP_001012026.1 | 3 - 4 | 261 | 116 | |
XM_006249777.2 | XP_006249839.1 | 3 - 4 | 322 | 116 | ||
XM_006249778.2 | XP_006249840.1 | 3 - 4 | 326 | 116 | ||
XM_006249779.2 | XP_006249841.1 | 3 - 4 | 323 | 116 | ||
GenBank mRNA | BC083647.1 | - | 3 - 4 | 261 | 7135053 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
