Assay Details
Gene Symbol: Ube2t | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108344.2 | NP_001101814.2 | 2 - 3 | 266 | 118 | |
GenBank mRNA | AW141189.1 | - | 2 - 3 | 266 | 118 | |
BC167012.1 | - | 2 - 3 | 227 | 9099623 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC167012.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
