Assay Details
Gene Symbol: Acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011906.1 | NP_001011906.1 | 6 - 7 | 1156 | 70 | |
XM_006250031.2 | XP_006250093.1 | 4 - 5 | 1023 | 70 | ||
GenBank mRNA | BC086598.1 | - | 6 - 7 | 1156 | 7313105 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P780943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25895 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P780943 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P780943 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P780943 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P780943 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103657 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80613 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46139 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89345 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15330 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7670 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7839 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93300 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48297 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44052 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10764819 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764815 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764817 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764818 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764820 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10764814 | - | - |
Rat Genome 230 2.0 Array | 1374555_at | - | - |
Rat Genome 230 2.0 Array | 1374555_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374555_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1374555_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300003679.rn.1 | TC1300000533.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300003680.rn.1 | TC1300000533.rn.1 | RTA TC |
Rat Expression Set 230 | 1374555_at | - | - |
Rat Expression Set 230 | 1374555_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10764814 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21917233 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376438 | - | - |
