Assay Details
Gene Symbol: Rgs16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250010.2 | XP_006250072.1 | 2 - 3 | 959 | 65 | |
XM_006250011.2 | XP_006250073.1 | 2 - 3 | 964 | 65 | ||
XM_006250012.1 | XP_006250074.1 | 2 - 3 | 1025 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
