Assay Details
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019344.2 | NP_062217.1 | 5 - 6 | 605 | 84 | |
XM_006250015.2 | XP_006250077.1 | 4 - 5 | 476 | 84 | ||
XM_006250016.2 | XP_006250078.1 | 5 - 6 | 420 | 84 | ||
XM_006250017.2 | XP_006250079.1 | 5 - 6 | 770 | 84 | ||
XM_008769675.1 | XP_008767897.1 | 5 - 6 | 771 | 84 | ||
XM_008769676.1 | XP_008767898.1 | 5 - 6 | 936 | 84 | ||
GenBank mRNA | AB006013.1 | - | 4 - 5 | 514 | 84 | |
BC089064.1 | - | 5 - 6 | 605 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
