Assay Details
Gene Symbol: Rab27b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254930.2 | XP_006254992.1 | 1 - 2 | 93 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006254930.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
