Assay Details
Gene Symbol: Ndst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024361.1 | NP_077337.1 | 9 - 10 | 2299 | 71 | |
XM_008772130.1 | XP_008770352.1 | 5 - 6 | 687 | 71 | ||
GenBank mRNA | M92042.1 | - | 9 - 10 | 2299 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
