Assay Details
Gene Symbol: Poli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106137.1 | NP_001099607.1 | 3 - 4 | 419 | 112 | |
XM_006254901.2 | XP_006254963.1 | 2 - 3 | 282 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
