Assay Details
Gene Symbol: Cd74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013069.2 | NP_037201.1 | 2 - 3 | 341 | 90 | |
XM_006254761.2 | XP_006254823.1 | 2 - 3 | 399 | 90 | ||
GenBank mRNA | BC059152.1 | - | 2 - 3 | 322 | 90 | |
FQ219430.1 | - | 2 - 3 | 90 | |||
FQ225098.1 | - | 2 - 3 | 316 | 90 | ||
FQ227357.1 | - | 2 - 3 | 318 | 90 | ||
FQ227455.1 | - | 2 - 3 | 321 | 90 | ||
FQ227561.1 | - | 2 - 3 | 323 | 90 | ||
FQ228116.1 | - | 2 - 3 | 324 | 90 | ||
FQ231756.1 | - | 2 - 3 | 325 | 90 | ||
FQ232412.1 | - | 2 - 3 | 366 | 90 | ||
FQ232423.1 | - | 2 - 3 | 355 | 90 | ||
FQ232867.1 | - | 2 - 3 | 325 | 90 | ||
FQ232996.1 | - | 2 - 3 | 318 | 90 | ||
FQ233568.1 | - | 2 - 3 | 324 | 90 | ||
FQ233706.1 | - | 2 - 3 | 324 | 90 | ||
FQ234457.1 | - | 2 - 3 | 327 | 90 | ||
X13044.1 | - | 2 - 3 | 297 | 90 | ||
X14254.1 | - | 2 - 3 | 305 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
