Assay Details
Gene Symbol: Cd74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013069.2 | NP_037201.1 | 3 - 4 | 425 | 69 | |
XM_006254761.2 | XP_006254823.1 | 3 - 4 | 483 | 69 | ||
GenBank mRNA | BC059152.1 | - | 3 - 4 | 406 | 69 | |
FQ225098.1 | - | 3 - 4 | 400 | 69 | ||
FQ225666.1 | - | 1 - 2 | 44 | 69 | ||
FQ227357.1 | - | 3 - 4 | 402 | 69 | ||
FQ227455.1 | - | 3 - 4 | 405 | 69 | ||
FQ227561.1 | - | 3 - 4 | 407 | 69 | ||
FQ228116.1 | - | 3 - 4 | 408 | 69 | ||
FQ231756.1 | - | 3 - 4 | 409 | 69 | ||
FQ232412.1 | - | 3 - 4 | 450 | 69 | ||
FQ232423.1 | - | 3 - 4 | 439 | 69 | ||
FQ232867.1 | - | 3 - 4 | 409 | 69 | ||
FQ232996.1 | - | 3 - 4 | 402 | 69 | ||
FQ233568.1 | - | 3 - 4 | 408 | 69 | ||
FQ233706.1 | - | 3 - 4 | 408 | 69 | ||
FQ234457.1 | - | 3 - 4 | 411 | 69 | ||
X13044.1 | - | 3 - 4 | 381 | 69 | ||
X14254.1 | - | 3 - 4 | 389 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
