Assay Details
Gene Symbol: Cd74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013069.2 | NP_037201.1 | 4 - 5 | 488 | 118 | |
XM_006254761.2 | XP_006254823.1 | 4 - 5 | 546 | 118 | ||
GenBank mRNA | BC059152.1 | - | 4 - 5 | 469 | 6890709 | 118 |
FQ225098.1 | - | 4 - 5 | 463 | 118 | ||
FQ225666.1 | - | 2 - 3 | 107 | 118 | ||
FQ227357.1 | - | 4 - 5 | 465 | 118 | ||
FQ227455.1 | - | 4 - 5 | 468 | 118 | ||
FQ228116.1 | - | 4 - 5 | 471 | 118 | ||
FQ232412.1 | - | 4 - 5 | 513 | 118 | ||
FQ232423.1 | - | 4 - 5 | 502 | 118 | ||
FQ232867.1 | - | 4 - 5 | 472 | 118 | ||
FQ232996.1 | - | 4 - 5 | 465 | 118 | ||
FQ233568.1 | - | 4 - 5 | 471 | 118 | ||
FQ233706.1 | - | 4 - 5 | 471 | 118 | ||
FQ234457.1 | - | 4 - 5 | 474 | 118 | ||
X13044.1 | - | 4 - 5 | 444 | 118 | ||
X14254.1 | - | 4 - 5 | 452 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
