Assay Details
Gene Symbol: Mbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115025.1 | NP_001108497.1 | 4 - 5 | 937 | 118 | |
GenBank mRNA | BC158877.1 | - | 4 - 5 | 937 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
