Assay Details
Gene Symbol: Snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008364.1 | NP_001008365.1 | 1 - 2 | 470 | 95 | |
XM_006254728.1 | XP_006254790.1 | 1 - 2 | 225 | 95 | ||
GenBank mRNA | BC085883.1 | - | 1 - 2 | 470 | 7192093 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
