Assay Details
Gene Symbol: Snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008364.1 | NP_001008365.1 | 6 - 7 | 845 | 113 | |
XM_008772152.1 | XP_008770374.1 | 7 - 8 | 892 | 113 | ||
GenBank mRNA | BC085883.1 | - | 6 - 7 | 845 | 7192093 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P897023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169685 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P897023 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P897023 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P897023 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P182879 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P897023 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19606 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79087 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78974 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49143 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12598 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53957 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48324 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96656 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76508 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15430 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57794 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72600 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10801757 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801756 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801753 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801755 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801754 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801758 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801760 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10801752 | - | - |
Rat Genome 230 2.0 Array | 1377603_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377603_at | - | - |
Affymetrix HT Rat Focus Array | 1377603_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377603_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800003833.rn.1 | TC1800000515.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800003832.rn.1 | TC1800000515.rn.1 | RTA TC |
Rat Expression Set 230 | 1377603_PM_at | - | - |
Rat Expression Set 230 | 1377603_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801752 | - | - |
Rat Genome U34 Set | rc_AI176764_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22191717 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365628 | - | - |
RatRef-12 Expression BeadChip | ILMN_69731 | - | - |
