Assay Details
Gene Symbol: Lmnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053905.1 | NP_446357.1 | 8 - 9 | 1817 | 61 | |
GenBank mRNA | U72353.1 | - | 8 - 9 | 1817 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
