Assay Details
Gene Symbol: Lmnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053905.1 | NP_446357.1 | 9 - 10 | 1939 | 88 | |
GenBank mRNA | U72353.1 | - | 9 - 10 | 1939 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P075647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30588 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P075647 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P075647 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P075647 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P325522 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57599 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79018 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100534 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10801829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801828 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10801821 | - | - |
Rat Genome 230 2.0 Array | 1373897_at | - | - |
Rat Genome 230 2.0 Array | 1368812_at | - | - |
Rat Genome 230 2.0 Array | 1373897_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368811_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368811_at | - | - |
Rat Genome 230 2.0 Array | 1368812_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373897_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368812_at | - | - |
Affymetrix HT Rat Focus Array | 1368812_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368811_at | - | - |
Affymetrix HT Rat Focus Array | 1368811_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373897_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800004045.rn.1 | TC1800000555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800004044.rn.1 | TC1800000555.rn.1 | RTA TC |
Rat Expression Set 230 | 1373897_PM_at | - | - |
Rat Expression Set 230 | 1368812_PM_at | - | - |
Rat Expression Set 230 | 1368811_at | - | - |
Rat Expression Set 230 | 1368812_at | - | - |
Rat Expression Set 230 | 1368811_PM_at | - | - |
Rat Expression Set 230 | 1373897_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801821 | - | - |
Rat Genome U34 Set | rc_AA957201_s_at | - | - |
Rat Genome U34 Set | U72353_at | - | - |
Rat Genome U34 Set | rc_AI103106_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21505339 | - | - |
