Assay Details
Gene Symbol: Lox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017061.2 | NP_058757.1 | 3 - 4 | 929 | 65 | |
XM_006254713.2 | XP_006254775.1 | 1 - 2 | 1012 | 65 | ||
XM_006254714.2 | XP_006254776.1 | 2 - 3 | 912 | 65 | ||
XM_006254715.2 | XP_006254777.1 | 1 - 2 | 1013 | 65 | ||
GenBank mRNA | BC078861.1 | - | 3 - 4 | 929 | 7113042 | 65 |
FQ230374.1 | - | 1 - 2 | 164 | 65 | ||
U11038.1 | - | 1 - 2 | 897 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
