Assay Details
Gene Symbol: Stard6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007627.1 | NP_001007628.1 | 1 - 2 | 141 | 100 | |
XM_006254921.2 | XP_006254983.1 | 2 - 3 | 363 | 100 | ||
XM_008772111.1 | XP_008770333.1 | 2 - 3 | 211 | 100 | ||
GenBank mRNA | AY555189.1 | - | 2 - 3 | 311 | 100 | |
BC078976.1 | - | 1 - 2 | 141 | 7111945 | 100 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY555189.1,BC078976.1,NM_001007627.1,XM_006254921.2,XM_008772111.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
