Assay Details
Gene Symbol: Apcdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001071384.4 | XP_001071384.2 | 1 - 2 | 137 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
