Assay Details
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145366.1 | NP_001138838.1 | 5 - 6 | 735 | 113 | |
NM_001145367.1 | NP_001138839.1 | 5 - 6 | 913 | 113 | ||
NM_013124.3 | NP_037256.1 | 4 - 5 | 710 | 113 | ||
XM_006237009.2 | XP_006237071.1 | 5 - 6 | 706 | 113 | ||
GenBank mRNA | AB011365.1 | - | 3 - 4 | 543 | 113 | |
AB019561.1 | - | 4 - 5 | 624 | 113 | ||
AF156665.1 | - | 5 - 6 | 735 | 113 | ||
AF156666.1 | - | 4 - 5 | 672 | 113 | ||
AF246457.1 | - | 5 - 6 | 663 | 113 | ||
AF246458.1 | - | 5 - 6 | 703 | 113 | ||
Y12882.2 | - | 4 - 5 | 673 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
