Assay Details
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145366.1 | NP_001138838.1 | 6 - 7 | 936 | 164 | |
NM_001145367.1 | NP_001138839.1 | 6 - 7 | 1114 | 164 | ||
NM_013124.3 | NP_037256.1 | 5 - 6 | 911 | 164 | ||
XM_006237009.2 | XP_006237071.1 | 6 - 7 | 907 | 164 | ||
GenBank mRNA | AB011365.1 | - | 4 - 5 | 744 | 164 | |
AB019561.1 | - | 5 - 6 | 825 | 164 | ||
AF156665.1 | - | 6 - 7 | 936 | 164 | ||
AF156666.1 | - | 5 - 6 | 873 | 164 | ||
AF246457.1 | - | 6 - 7 | 864 | 164 | ||
AF246458.1 | - | 6 - 7 | 904 | 164 | ||
Y12882.2 | - | 5 - 6 | 874 | 164 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
