Assay Details
Gene Symbol: Pparg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145366.1 | NP_001138838.1 | 1 - 2 | 120 | 132 | |
GenBank mRNA | AF156665.1 | - | 1 - 2 | 120 | 132 | |
AF246457.1 | - | 1 - 2 | 48 | 132 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF156665.1,AF246457.1,NM_001145366.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
