Assay Details
Gene Symbol: Tm7sf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011970.1 | NP_001011970.1 | 8 - 9 | 1123 | 64 | |
GenBank mRNA | BC090030.1 | - | 8 - 9 | 1123 | 7366240 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
