Assay Details
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107885.1 | NP_001101355.1 | 1 - 2 | 136 | 73 | |
XM_006237248.1 | XP_006237310.1 | 1 - 2 | 140 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001107885.1, XM_006237248.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
