Assay Details
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107885.1 | NP_001101355.1 | 3 - 4 | 417 | 148 | |
XM_006237248.1 | XP_006237310.1 | 3 - 4 | 421 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
