Assay Details
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107885.1 | NP_001101355.1 | 6 - 7 | 778 | 71 | |
XM_006237248.1 | XP_006237310.1 | 6 - 7 | 796 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P18010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29061 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18010 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18010 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P18010 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17344 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4476 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15872 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10858364 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10858366 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10858368 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10702442 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10858363 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10858367 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10858365 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10702441 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10858362 | - | - |
Rat Genome 230 2.0 Array | 1372279_at | - | - |
Rat Genome 230 2.0 Array | 1372279_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372279_at | - | - |
Affymetrix HT Rat Focus Array | 1372279_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400012123.rn.1 | TC0400001695.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0400012124.rn.1 | TC0400001695.rn.1 | RTA TC |
Rat Expression Set 230 | 1372279_PM_at | - | - |
Rat Expression Set 230 | 1372279_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10858362 | - | - |
Rat Genome U34 Set | rc_AA892271_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21435551 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369596 | - | - |
