Assay Details
Gene Symbol: Ccnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | D16308.1 | - | 4 - 5 | 895 | 57 | |
FQ227079.1 | - | 4 - 5 | 927 | 57 | ||
FQ233117.1 | - | 4 - 5 | 927 | 57 | ||
FQ234359.1 | - | 4 - 5 | 927 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
