Assay Details
Gene Symbol: Usp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014058.1 | NP_001014080.1 | 6 - 7 | 827 | 166 | |
GenBank mRNA | BC079101.1 | - | 6 - 7 | 827 | 7124857 | 166 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
