Assay Details
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763442.1 | XP_008761664.1 | 2 - 3 | 361 | 78 | |
XM_008763443.1 | XP_008761665.1 | 3 - 4 | 1116 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
