Assay Details
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763441.1 | XP_008761663.1 | 4 - 5 | 1045 | 108 | |
XM_008763442.1 | XP_008761664.1 | 6 - 7 | 1033 | 108 | ||
XM_008763443.1 | XP_008761665.1 | 7 - 8 | 1788 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
