Assay Details
Gene Symbol: Bcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017253.2 | NP_058949.1 | 5 - 6 | 684 | 82 | |
XM_008763370.1 | XP_008761592.1 | 5 - 6 | 697 | 82 | ||
XM_008763371.1 | XP_008761593.1 | 5 - 6 | 947 | 82 | ||
XM_008763372.1 | XP_008761594.1 | 6 - 7 | 773 | 82 | ||
XM_008763373.1 | XP_008761595.1 | 5 - 6 | 876 | 82 | ||
GenBank mRNA | AF165887.1 | - | 3 - 4 | 441 | 82 | |
AJ278701.1 | - | 5 - 6 | 678 | 82 | ||
BC087710.1 | - | 5 - 6 | 684 | 7315507 | 82 | |
U35774.1 | - | 5 - 6 | 650 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
