Assay Details
Gene Symbol: Bcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017253.2 | NP_058949.1 | 9 - 10 | 1218 | 85 | |
XM_008763370.1 | XP_008761592.1 | 9 - 10 | 1231 | 85 | ||
XM_008763371.1 | XP_008761593.1 | 9 - 10 | 1481 | 85 | ||
XM_008763372.1 | XP_008761594.1 | 10 - 11 | 1307 | 85 | ||
XM_008763373.1 | XP_008761595.1 | 9 - 10 | 1410 | 85 | ||
GenBank mRNA | AF165887.1 | - | 7 - 8 | 975 | 85 | |
AJ278701.1 | - | 9 - 10 | 1212 | 85 | ||
BC087710.1 | - | 9 - 10 | 1218 | 85 | ||
U35774.1 | - | 9 - 10 | 1184 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
