Assay Details
Gene Symbol: Rho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033441.1 | NP_254276.1 | 2 - 3 | 612 | 145 | |
GenBank mRNA | Z46957.1 | - | 2 - 3 | 612 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
