Assay Details
Gene Symbol: Ccdc77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191752.1 | NP_001178681.1 | 2 - 3 | 277 | 94 | |
XM_006237236.2 | XP_006237298.1 | 3 - 4 | 523 | 94 | ||
XM_006237237.2 | XP_006237299.1 | 3 - 4 | 511 | 94 | ||
XM_006237238.1 | XP_006237300.1 | 4 - 5 | 566 | 94 | ||
XM_006237239.1 | XP_006237301.1 | 4 - 5 | 548 | 94 | ||
XM_006237240.2 | XP_006237302.1 | 3 - 4 | 1434 | 94 | ||
XM_006237241.1 | XP_006237303.1 | 3 - 4 | 369 | 94 | ||
XM_006237242.2 | XP_006237304.1 | 2 - 3 | 1454 | 94 | ||
XM_006237243.2 | XP_006237305.1 | 2 - 3 | 1380 | 94 | ||
XM_006237244.2 | XP_006237306.1 | 2 - 3 | 277 | 94 | ||
XM_008763244.1 | XP_008761466.1 | 3 - 4 | 523 | 94 | ||
XM_008763245.1 | XP_008761467.1 | 3 - 4 | 1508 | 94 | ||
XM_008763246.1 | XP_008761468.1 | 3 - 4 | 364 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
